missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D-amino-acid oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) D-amino-acid oxidase, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001908.3 |
---|---|
Gene ID (Entrez) | 1610 |
Name | D-amino-acid oxidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958609
|
enQuireBio™
QP5910-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998609
|
enQuireBio™
QP5910-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968609
|
enQuireBio™
QP5910-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988609
|
enQuireBio™
QP5910-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908619
|
enQuireBio™
QP5910-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978609
|
enQuireBio™
QP5910-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001908.3 | |
D-amino-acid oxidase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1610 | |
Research Use Only | |
DAO | |
Recombinant Protein | |
Yeast | |
MRVVVIGAGV IGLSTALCIH ERYHSVLQPL DIKVYADRFT PLTTTDVAAG LWQPYLSDPN NPQEADWSQQ TFDYLLSHVH SPNAENLGLF LISGYNLFHE AIPDPSWKDT VLGFRKLTPR ELDMFPDYGY GWFHTSLILE GKNYLQWLTE RLTERGVKFF QRKVESFEEV AREGADVIVN CTGVWAGALQ RDPLLQPGRG QIMKVDAPWM KHFILTHDPE RGIYNSPYII PGTQTVTLGG IFQLGNWSEL NNIQDHNTIW EGCCRLEPTL KNARIIGERT GFRPVRPQIR LEREQLRTGP SNTEVIHNYG HGGYGLTIHW GCALEAAKLF GRILEEKKLS RMPPSHL | |
Greater than 90% as determined by SDS-PAGE. |