missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D-glucuronyl C5-epimerase Protein
A DNA sequence encoding the Homo sapiens (Human) D-glucuronyl C5-epimerase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6087-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001311022.1 | |
D-glucuronyl C5-epimerase Protein | |
Research Use Only | |
GLCE | |
Recombinant Protein | |
E. coli | |
NKCSSDKAIQ FPRRSSSGFR VDGFEKRAAA SESNNYMNHV AKQQSEEAFP QEQQKAPPVV GGFNSNVGSK VLGLKYEEID CLINDEHTIK GRREGNEVFL PFTWVEKYFD VYGKVVQYDG YDRFEFSHSY SKVYAQRAPY HPDGVFMSFE GYNVEVRDRV KCISGVEGVP LSTQWGPQGY FYPIQIAQYG LSHYSKNLTE KPPHIEVYET AEDRDKNKPN DWTVPKGCFM ANVADKSRFT NVKQFIAPET SEGVSLQLGN TKDFIISFDL KFLTNGSVSV VLETTEKNQL FTIHYVSNAQ LIAFKERDIY YGIGPRTSWS TVTRDLVTDL RKGVGLSNTK AVKPTKIMPK KVVRLIAKGK GFLDNITIST TAHMAAFFAA SDWLVRNQDE KGGWPIMVTR KLGEGFKSLE PGWYSAMAQG QAISTLVRAY LLTKDHIFLN SALRATAPYK FLSEQHGVKA VFMNKHDWYE EYPTTPSSFV LNGFMYSLIG LYDLKETAGE KLGKEARSLY ERGMESLKAM LPLYDTGSGT IYDLRHFMLG IAPNLARWDY HTTHINQLQL LSTIDESPVF KEFVKRWKSY LKGSRAKHN | |
Greater than 90% as determined by SDS-PAGE. |
26035 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |