missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Defensin beta 2 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-34910-100ug
This item is not returnable.
View return policy
Description
Defensin beta 2 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using immature human dendritic cells is in a concentration range of 10-100 ng/mL.Specifications
O15263, O15263, O15263 | |
PAGE, Bioactivity | |
13215 | |
100 μg | |
Less than 1 EU/ug of Defensin beta 2 as determined by LAL method. | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>98% pure by SDS-PAGE and HPLC |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. | |
4.3 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
BD-2, Defb2, Defb2 defensin beta 2, defensin beta 2 | |
Biologically Active Proteins | |
Defensin beta 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction