missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Density-regulated Protein
A DNA sequence encoding the Homo sapiens (Human) Density-regulated protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5928-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_003668.2 | |
Density-regulated Protein | |
Research Use Only | |
DENR | |
Recombinant Protein | |
E. coli | |
AADISESSGA DCKGDPRNSA KLDADYPLRV LYCGVCSLPT EYCEYMPDVA KCRQWLEKNF PNEFAKLTVE NSPKQEAGIS EGQGTAGEEE EKKKQKRGGR GQIKQKKKTV PQKVTIAKIP RAKKKYVTRV CGLATFEIDL KEAQRFFAQK FSCGASVTGE DEIIIQGDFT DDIIDVIQEK WPEVDDDSIE DLGEVKK | |
Greater than 90% as determined by SDS-PAGE. |
8562 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |