missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Dkk-4 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

£422.00

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 27121
Molecular Weight (g/mol) 24.4 kDa
Name Dkk-4
Quantity 10 μg
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18117590
View Documents Promotion Details
R&D Systems™
1269-DK-010/CF
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
£422.00
10µg
Please to purchase this item. Need a web account? Register with us today!
18146810
View Documents Promotion Details
R&D Systems™
1269-DK-010
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein.
£422.00
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human Dkk-4 Protein is derived from NS0. The Recombinant Human Dkk-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Bioactivity
24.4 kDa
10 μg
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
27121
Dkk-4
Mouse myeloma cell line,NS0-derived human Dkk-4 protein Leu19-Leu224,with a C-terminal 10-His tag
Recombinant
Videos
SDS
Documents

Documents

Product Certifications