missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Docking protein 1
A DNA sequence encoding the Homo sapiens (Human) Docking protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5943-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_060901.2 | |
Docking protein 1 | |
Research Use Only | |
DOK5 | |
Recombinant Protein | |
E. coli | |
MECVGTRIND ISLGEPDLLA TGVEREQSER FNVYLMPSPN LDVHGECALQ ITYEYICLWD VQNPRVKLIS WPLSALRRYG RDTTWFTFEA GRMCETGEGL FIFQTRDGEA IYQKVHSAAL AIAEQHERLL QSVKNSMLQM KMSERAASLS TMVPLPRSAY WQHITRQHST GQLYRLQDVS SPLKLHRTET FPAYRSEH | |
Greater than 90% as determined by SDS-PAGE. |
55816 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |