missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RNF125 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RNF125, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7870-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060301.2 | |
E3 ubiquitin-protein ligase RNF125 Protein | |
Research Use Only | |
RNF125 | |
Recombinant Protein | |
E. coli | |
GSVLSTDSGK SAPASATARA LERRRDPELP VTSFDCAVCL EVLHQPVRTR CGHVFCRSCI ATSLKNNKWT CPYCRAYLPS EGVPATDVAK RMKSEYKNCA ECDTLVCLSE MRAHIRTCQK YIDKYGPLQE LEETAARCVC PFCQRELYED SLLDHCITHH RSERRPVFCP LCRLIPDENP SSFSGSLIRH LQVSHTLFYD DFIDFNIIEE ALIRRVLDRS LLEYVNHSNT T | |
Greater than 80% as determined by SDS-PAGE. |
54941 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |