missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF1AX Protein

A cDNA sequence encoding the EIF1AX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11763-5ug

 View more versions of this product

Product Code. 15968288

  • £99.15 / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

EIF1AX Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPKNKGKGGK NRRRGKNENE SEKRELVFKE DGQEYAQVIK MLGNGRLEAM CFDGVKRLCH IRGKLRKKVW INTSDIILVG LRDYQDNKAD VILKYNADEA RSLKAYGELP EHAKINETDT FGPGDDDEIQ FDDIGDDDED IDDI
Greater than 90.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EIF1AX protein solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 5mM DTT, 200mM NaCl and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers