missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endoplasmic reticulum resident protein 44 Protein
A DNA sequence encoding the Homo sapiens (Human) Endoplasmic reticulum resident protein 44, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8042-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055866.1 | |
Endoplasmic reticulum resident protein 44 Protein | |
Research Use Only | |
ERP44 | |
Recombinant Protein | |
E. coli | |
EITSLDTENI DEILNNADVA LVNFYADWCR FSQMLHPIFE EASDVIKEEF PNENQVVFAR VDCDQHSDIA QRYRISKYPT LKLFRNGMMM KREYRGQRSV KALADYIRQQ KSDPIQEIRD LAEITTLDRS KRNIIGYFEQ KDSDNYRVFE RVANILHDDC AFLSAFGDVS KPERYSGDNI IYKPPGHSAP DMVYLGAMTN FDVTYNWIQD KCVPLVREIT FENGEELTEE GLPFLILFHM KEDTESLEIF QNEVARQLIS EKGTINFLHA DCDKFRHPLL HIQKTPADCP VIAIDSFRHM YVFGDFKDVL IPGKLKQFVF DLHSGKLHRE FHHGPDPTDT APGEQAQDVA SSPPESSFQK LAPSEYRYTL LRDRDEL | |
Greater than 90% as determined by SDS-PAGE. |
23071 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |