missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ephrin A4 Protein
A cDNA sequence encoding the Ephrin A4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11800-5ug
This item is not returnable.
View return policy
Specifications
1945 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRLLPLLRTV LWAAFLGSPL RGGSSLRHVV YWNSSNPRLL RGDAVVELGL NDYLDIVCPH YEGPGPPEGP ETFALYMVDW PGYESCQAEG PRAYKRWVCS LPFGHVQFSE KIQRFTPFSL GFEFLPGETY YYISVPTPES SGQCLRLQVS VCCKERKSES AHPVGSPGES GTSGWRGGDT PSPLCLLLLL LLLILRLLRI L |
Ephrin A4 Protein | |
Research Use Only | |
EFNA4 | |
Human | |
GST | |
Ephrin A4 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |