missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Excitatory amino acid transporter 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6690-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035140

  • £1735.00 / 1mg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

NP_004161.4
Excitatory amino acid transporter 3 Protein
Research Use Only
SLC1A1
Recombinant Protein
E. coli
AEDVTLIIAV DWLLDRFRTM VNVLGDAFGT GIVEKLSKKE LEQMDVSSEV NIVNPFALES TILDNEDSDT KKSYVNGGFA VDKSDTISFT QTSQF
Greater than 90% as determined by SDS-PAGE.
6505
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers