missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Excitatory amino acid transporter 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6692-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095150

  • £476.00 / 100µg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001259016.1
Excitatory amino acid transporter 4 Protein
Research Use Only
SLC1A6
Recombinant Protein
E. coli
MVTIIHPGKG SKEGLHREGR IETIPTADAF MDLIRNMFPP NLVEACFKQF KTQYSTRVVT RTMVRTENGS EPGASMPPPF SVENGTSFLE NVTRALGTLQ EMLSFEETVP VPGSANGINA LGL
Greater than 90% as determined by SDS-PAGE.
6511
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers