missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F-box / LRR-repeat protein 18 Protein

A DNA sequence encoding the Homo sapiens (Human) F-box/LRR-repeat protein 18, was expressed in the hosts and tags indicated.

£145.46 - £1735.00

Specifications

Name F-box / LRR-repeat protein 18 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol FBXL18
Protein Length Partial (see sequence information for more details). of Isoform 3
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15959429
View Documents Promotion Details
enQuireBio™
QP6024-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15999429
View Documents Promotion Details
enQuireBio™
QP6024-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15949429
View Documents Promotion Details
enQuireBio™
QP6024-ec-100ug
100 μg
£384.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15979429
View Documents Promotion Details
enQuireBio™
QP6024-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15989429
View Documents Promotion Details
enQuireBio™
QP6024-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15969429
View Documents Promotion Details
enQuireBio™
QP6024-ec-1mg
1 mg
£1735.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

F-box / LRR-repeat protein 18 Protein
Not determined.
Partial (see sequence information for more details). of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
FBXL18
Recombinant Protein
E. coli
MASSGEDISN DDDDMHPAAA GMADGVHLLG FSDEILLHIL SHVPSTDLIL NVRRTCRKLA ALCLDKSLIH TVLLQKDYQA SEDKVRQLVK EIGREIQQLS MAGCYWLPGS TVEHVARCRS LVKVNLSGCH LTSLRLSKML SALQHLRSLA IDVSPGFDAS QLSSECKATL SRVRELKQTL FTPSYGVVPC CTSLEKLLLY FEILDRTREG AILSGQLMVG QSNVPHYQNL RVFYARLAPG YINQEVVRLY LAVLSDRTPQ NLHAFLISVP GSFAESGATK NLLDSMARNV VLDALQLPKS WLNGSSLLQH MKFNNPFYFS FSRCTLSGGH LIQQVINGGK DLRSLASLNL SGCVHCLSPD SLLCR
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications