missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fas Ligand / FASLG / CD95L Protein
A DNA sequence encoding the Homo sapiens (Human) Fas Ligand / FASLG / CD95L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6021-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_000630.1 | |
Fas Ligand / FASLG / CD95L Protein | |
Research Use Only | |
FASLG | |
Recombinant Protein | |
Yeast | |
QIGHPSPPPE KKELRKVAHL TGKSNSRSMP LEWEDTYGIV LLSGVKYKKG GLVINETGLY FVYSKVYFRG QSCNNLPLSH KVYMRNSKYP QDLVMMEGKM MSYCTTGQMW ARSSYLGAVF NLTSADHLYV NVSELSLVNF EESQTFFGLY KL | |
Greater than 90% as determined by SDS-PAGE. |
356 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |