missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIIA/CD16a Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 4325-FC-050

Additional Details : Weight : 0.00970kg

Product Code. 18188160

  • £387.68 / 50µg
Estimated Shipment: 24-07-2024
to see stock.

Description

Description

The Recombinant Human Fc gamma RIIIA/CD16a Protein is derived from NS0. The Recombinant Human Fc gamma RIIIA/CD16a Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22.6 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
2214
Fc gamma RIIIA/CD16a
Mouse myeloma cell line,NS0-derived human Fc gamma RIIIA/CD16a protein Gly17-Gln208,with a C-terminal 6-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers