missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCGR3A Protein
A cDNA sequence encoding the FCGR3A was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 2214 |
---|---|
Name | FCGR3A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | FCGR3A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968608
|
enQuireBio™
QP11860-HIS-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15978608
|
enQuireBio™
QP11860-HIS-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15948608
|
enQuireBio™
QP11860-HIS-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958608
|
enQuireBio™
QP11860-HIS-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15988608
|
enQuireBio™
QP11860-HIS-EC-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
2214 | |
Research Use Only | |
FCGR3A | |
Human | |
His | |
FCGR3A protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 10% glycerol. |
FCGR3A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMRT EDLPKAVVFL EPQWYRVLEK DSVTLKCQGA YSPEDNSTQW FHNESLISSQ ASSYFIDAAT VDDSGEYRCQ TNLSTLSDPV QLEVHIGWLL LQAPRWVFKE EDPIHLRCHS WKNTALHKVT YLQNGKGRKY FHHNSDFYIP KATLKDSGSY FCRGLFGSKN VSSETVNITI TQGLAVSTIS SFFPPGYQ | |
Greater than 90% as determined by SDS-PAGE. |