missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FEN1 Protein
A cDNA sequence encoding the FEN1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Gene ID (Entrez) | 2237 |
---|---|
Name | FEN1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | FEN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978628
|
enQuireBio™
QP11866-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968628
|
enQuireBio™
QP11866-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958628
|
enQuireBio™
QP11866-1mg |
1 mg |
£3965.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
2237 | |
Research Use Only | |
FEN1 | |
Human | |
Untagged | |
The protein contains 20mM Tris-HCl buffer pH 8.0, 1mM DTT, 10% glycerol. |
FEN1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGIQGLAKLI ADVAPSAIRE NDIKSYFGRK VAIDASMSIY QFLIAVRQGG DVLQNEEGET TSHLMGMFYR TIRMMENGIK PVYVFDGKPP QLKSGELAKR SERRAEAEKQ LQQAQAAGAE QEVEKFTKRL VKVTKQHNDE CKHLLSLMGI PYLDAPSEAE ASCAALVKAG KVYAAATEDM DCLTFGSPVL MRHLTASEAK KLPIQEFHLS RILQELGLNQ EQFVDLCILL GSDYCESIRG IGPKRAVDLI QKHKSIEEIV RRLDPNKYPV PENWLHKEAH QLFLEPEVLD PESVELKWSE PNEEELIKFM CGEKQFSEER IRSGVKRLSK SRQGSTQGRL DDFFKVTGSL SSAKRKEPEP KGSTKKKAKT GAAGKFKRGK | |
Greater than 90.0% as determined by SDS-PAGE. |