missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-6 (aa 67-208) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£211.66
Specifications
Gene ID (Entrez) | 2251 |
---|---|
Molecular Weight (g/mol) | 16.1 kDa |
Name | FGF-6 (aa 67-208) |
Quantity | 25μg |
Source | E. coli-derived human FGF-6 protein Gly67-Ile208 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18128030
|
R&D Systems™
6829-F6-025 |
Lyophilized from a 0.2μm filtered solution in MES, Na2S04, NaCl and CHAPS with BSA as a carrier protein. |
£211.66
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18167390
|
R&D Systems™
6829-F6-025/CF |
Lyophilized from a 0.2μm filtered solution in MES, Na2S04, NaCl and CHAPS. |
£211.66
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human FGF-6 (aa 67-208) Protein is derived from E. coli. The Recombinant Human FGF-6 (aa 67-208) Protein has been validated for the following applications: Bioactivity.Specifications
2251 | |
FGF-6 (aa 67-208) | |
E. coli-derived human FGF-6 protein Gly67-Ile208 | |
Monomer | |
Recombinant |
16.1 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |