missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (157 aa) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 234-FSE-025/CF

247.08 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18156710

  • £323.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human FGF basic/FGF2 (157 aa) Protein is derived from E. coli. The Recombinant Human FGF basic/FGF2 (157 aa) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl.
17.4 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Recombinant
2247
FGF basic 157 aa
E. coli-derived human FGF basic/FGF2/bFGF protein Gly132-Ser288
<0.1 EU per 1μg of the protein by the LAL method.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers