missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR1 Protein
A cDNA sequence encoding the FGFR1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10604-10ug
This item is not returnable.
View return policy
Specifications
2260 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by its ability to inhibit human FGF acidic-dependent proliferation on R1 cells. The ED50 for this effect is typically at 15.0-30.0 ng/ml. | |
Human | |
Untagged | |
CD331 was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |
FGFR1 Protein | |
Research Use Only | |
FGFR1 | |
Recombinant Protein | |
Baculovirus | |
MWSWKCLLFW AVLVTATLCT ARPSPTLPEQ AQPWGAPVEV ESFLVHPGDL LQLRCRLRDD VQSINWLRDG VQLAESNRTR ITGEEVEVQD SVPADSGLYA CVTSSPSGSD TTYFSVNVSD ALPSSEDDDD DDDSSSEEKE TDNTKPNRMP VAPYWTSPEK MEKKLHAVPA AKTVKFKCPS SGTPNPTLRW LKNGKEFKPD HRIGGYKVRY ATWSIIMDSV VPSDKGNYTC IVENEYGSIN HTYQLDVVER SPHRPILQAG LPANKTVALG SNVEFMCKVY SDPQPHIQWL KHIEVNGSKI GPDNLPYVQI LKTAGVNTTD KEMEVLHLRN VSFEDAGEYT CLAGNSIGLS HHSAWLTVLE ALEERPAVMT SPLYLEIIIY CTGAFLISCM VGSVIVYKMK SGTKKSDFHS QMAVHKLAKS IPLRRQVTVS ADSSASMNSG VLLVRPSRLS SSGTPMLAGV SEYELPEDPR WELPRDRLVL GKPLGEGCFG QVVLAEAIGL DKDKPNRVTK VAVKMLKSDA TEKDLSDLIS EMEMMKMIGK HKNIINLLGA CTQDGPLYVI VEYASKGNLR EYLQARRPPG LEYCYNPSHN PEEQLSSKDL VSCAYQVARG MEYLASKKCI HRDLAARNVL VTEDNVMKIA DFGLARDIHH IDYYKKTTNG RLPVKWMAPE ALFDRIYTHQ SDVWSFGVLL WEIFTLGGSP YPGVPVEELF KLLKEGHRMD KPSNCTNELY MMMRDCWHAV PSQRPTFKQL VEDLDRIVAL TSNQEYLDLS MPLDQYSPSF PDTRSSTCSS GEDSVFSHEP LPEEPCLPRH PAQLANGGLK RR | |
Greater than 90.0% as determined by SDS-PAGE. |