missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR1OP Protein
A cDNA sequence encoding the FGFR1OP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11875-1mg
This item is not returnable.
View return policy
Specifications
11116 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAATAA AVVAEEDTEL RDLLVQTLEN SGVLNRIKAE LRAAVFLALE EQEKVENKTP LVNESLKKFL NTKDGRLVAS LVAEFLQFFN LDFTLAVFQP ETSTLQGLEG RENLARDLGI IEAEGTVGGP LLLEVIRRCQ QKEKGPTTGE GALDLSDVHS PPKSPEGKTS AQTTPSKKAN DEANQSDTSV SLSEPKSKSS LHLLSHETKI GSFLSNRTLD GKDKAGLCPD EDDMEGDSFF DDPIPKPEKT YGLRKEPRKQ AGSLASLSDA PPLKSGLSSL AGAPSLKDSE SKRGNTVLKD LKLISDKIGS LGLGTGEDDD YVDDFNSTSH RSEKSEISIG EEIEEDLSVE IDDINTSDKL DDLTQDLTVS QLSDVADYLE DVA | |
Greater than 85.0% as determined by SDS-PAGE. |
FGFR1OP Protein | |
Research Use Only | |
FGFR1OP | |
Human | |
His | |
FGFR1OP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |