missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Filaggrin Protein
A DNA sequence encoding the Homo sapiens (Human) Filaggrin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6050-ba-100ug
This item is not returnable.
View return policy
Specifications
NP_002007.1 | |
Filaggrin Protein | |
Research Use Only | |
FLG | |
Recombinant Protein | |
Baculovirus | |
DSSRHSQSGQ GESAGSRRSR RQGSSVSQDS DSEAYPEDSE RRSESASRNH HGSSREQSRD GSRHPGSSHR DTASHVQSSP VQSDSSTAKE HGHFSSLSQD SAYHSGIQSR GSPHSSSSYH YQSEGTERQK GQSGLVWRHG SYGSADYDYG ESGFRHSQHG SVSYNSNPVV FKERSDICKA SAFGKDHPRY YATYINKDPG LCGHSSDISK QLGFSQSQRY YYYE | |
Greater than 90% as determined by SDS-PAGE. |
2312 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |