missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FKBP12 / FKBP1A Protein
A DNA sequence encoding the Homo sapiens (Human) FKBP12 / FKBP1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8403-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_000792.1 | |
FKBP12 / FKBP1A Protein | |
Research Use Only | |
FKBP1A | |
Recombinant Protein | |
E. coli | |
GVQVETISPG DGRTFPKRGQ TCVVHYTGML EDGKKFDSSR DRNKPFKFML GKQEVIRGWE EGVAQMSVGQ RAKLTISPDY AYGATGHPGI IPPHATLVFD VE | |
Greater than 90% as determined by SDS-PAGE. |
2280 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |