missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Fc (HEK293) Protein

Brand:  R&D Systems™ 11309-FKH-250

1465.19 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18709176

  • £1860.00 / 250µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.300-4.80 ng/mL.
Specifications

Specifications

AAA17999.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W. Observed: 57-64 kDa, under reducing conditions., M.W. Theoretical: 45 kDa
Human embryonic kidney cell, HEK293-derived human Flt-3 Ligand/FLT3L protein Human Flt-3 Ligand/FLT3L (Thr27-Pro185) (N-terminus) Accession # AAA17999.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
<0.10 EU per 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Bioactivity
2323
250 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Flt-3 Ligand/FLT3L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers