missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FRZB Protein
A cDNA sequence encoding the FRZB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11900-20ug
This item is not returnable.
View return policy
Specifications
FRZB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAACEPVR IPLCKSLPWN MTKMPNHLHH STQANAILAI EQFEGLLGTH CSPDLLFFLC AMYAPICTID FQHEPIKPCK SVCERARQGC EPILIKYRHS WPENLACEEL PVYDRGVCIS PEAIVTADGA DFPMDSSNGN CRGASSERCK CKPIRATQKT YFRNNYNYVI RAKVKEIKTK CHDVTAVVEV KEILKSSLVN IPRDTVNLYT SSGCLCPPLN VNEEYIIMGY EDEERSRLLL VEGSIAEKWK DRLGKKVKRW DMKLRHLGLS KSDSSNSDST QSQKSGRNSN PRQARN | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FRZB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |