missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FSH alpha/beta Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

£370.00

Specifications

Gene ID (Entrez) 2488
Molecular Weight (g/mol) 10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
Name FSH alpha/beta
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human FSH alpha/beta protein, (N-terminus) Human FSH alpha Ala25-Ser116 Accession &Num; P01215 Human FSH beta Cys21-Glu129 Accession &Num; P01225 (C-terminus)
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18309467
View Documents
R&D Systems™
5925-FS-010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
£370.00
10µg
Please to purchase this item. Need a web account? Register with us today!
18328377
View Documents
R&D Systems™
5925-FS-010/CF
Lyophilized from a 0.2μm filtered solution in PBS.
£370.00
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human FSH alpha/beta Protein is derived from CHO. The Recombinant Human FSH alpha/beta Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

2488
FSH alpha/beta
Chinese Hamster Ovary cell line,CHO-derived human FSH alpha/beta protein, (N-terminus) Human FSH alpha Ala25-Ser116 Accession &Num; P01215 Human FSH beta Cys21-Glu129 Accession &Num; P01225 (C-terminus)
Noncovalently-linked heterodimer
Recombinant
10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE with silver staining.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.