missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FXYD5 Protein
A cDNA sequence encoding the FXYD5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11913-1mg
This item is not returnable.
View return policy
Specifications
FXYD5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQTLKDTT SSSSADSTIM DIQVPTRAPD AVYTELQPTS PTPTWPADET PQPQTQTQQL EGTDGPLVTD PETHKSTKAA HPTDDTTTLS ERPSPSTDVQ TDPQTLKPSG FHEDDPFFYD EHTLRKR | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FXYD5 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |