Learn More
R&D Systems™ Recombinant Human Galectin-2 (HEK-293-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 9874-GA-050
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18197890
  • £337.00 / 50µg
Explore more special offers
Description
Description
  • Derived from HEK293
  • Validated for the following applications: Bioactivity.
Specifications
Specifications
P05162
3957
50 μg
12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 400 μg/mL in water
> 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Bioactivity
12 to 15 kDa, reducing conditions
Human embryonic kidney cell, HEK293-derived human Galectin-2 protein Met1-Glu132
< 0.10 EU per 1 μg of the protein by the LAL method
Human
Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose