missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GATE-16 / GABARAPL2 Protein
A DNA sequence encoding the Homo sapiens (Human) GATE-16 / GABARAPL2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8448-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_009216.1 | |
GATE-16 / GABARAPL2 Protein | |
Research Use Only | |
GABARAPL2 | |
Recombinant Protein | |
E. coli | |
MKWMFKEDHS LEHRCVESAK IRAKYPDRVP VIVEKVSGSQ IVDIDKRKYL VPSDITVAQF MWIIRKRIQL PSEKAIFLFV DKTVPQSSLT MGQLYEKEKD EDGFLYVAYS GENTFGF | |
Greater than 90% as determined by SDS-PAGE. |
11345 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |