missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GCGR Protein

A cDNA sequence encoding the GCGR was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11939-2ug

 View more versions of this product

Product Code. 15978858

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

GCGR Protein
Research Use Only
Recombinant Protein
E. coli
MQVMDFLFEK WKLYGDQCHH NLSLLPPPTE LVCNRTFDKY SCWPDTPANT TANISCPWYL PWHHKVQHRF VFKRCGPDGQ WVRGPRGQPW RDASQCQMDG EEIEVQKEVA KMYSSFQLEH HHHHH
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GCGR filtered (0.4 microm) and lyophilized from solution in acetonitrile/0,1%TFA and 1% (w/v) trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers