missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-15 Protein
A DNA sequence encoding the Homo sapiens (Human) GDF-15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP4998-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_004855.2 | |
GDF-15 Protein | |
Research Use Only | |
GDF15 | |
Recombinant Protein | |
Yeast | |
RNGDHCPLGP GRCCRLHTVR ASLEDLGWAD WVLSPREVQV TMCIGACPSQ FRAANMHAQI KTSLHRLKPD TVPAPCCVPA SYNPMVLIQK TDTGVSLQTY DDLLAKDCHC I | |
Greater than 90% as determined by SDS-PAGE. |
9518 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |