missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF11 Protein

A DNA sequence encoding the Homo sapiens (Human) GDF11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6074-ec-100ug

 View more versions of this product

Product Code. 15949789

  • £384.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_005802.1
GDF11 Protein
Research Use Only
GDF11
Recombinant Protein
E. coli
NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
Greater than 90% as determined by SDS-PAGE.
10220
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers