missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GFPT1 Protein
A cDNA sequence encoding the GFPT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11956-1mg
This item is not returnable.
View return policy
Specifications
GFPT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQQIMKGN FSSFMQKEIF EQPESVVNTM RGRVNFDDYT VNLGGLKDHI KEIQRCRRLI LIACGTSYHA GVATRQVLEE LTELPVMVEL ASDFLDRNTP VFRDDVCFFL SQSGETADTL MGLRYCKERG ALTVGITNTV GSSISRETDC GVHINAGPEI GVASTKAYTS QFVSLVMFAL MMCDDRISMQ ERRKEIMLGL KRLPDLIKEV LSMDDEIQKL ATELYHQKSV LIMGRGYHYA TCLEGALKIK EITYMHSEGI LAGELKHGPL ALVDKLMPVI MIIMRDHTYA KCQNALQQVV ARQGRPVVIC DKEDTETIKN TKRTIKVPHS VDCLQGILSV IPLQLLAFHL AVLRGYDVDF PRNLAKSVTV E | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GFPT1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol, 2mM DTT and 2mM EDTA. |