missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucagon Protein
A DNA sequence encoding the Homo sapiens (Human) Glucagon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8639-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_002045.1 | |
Glucagon Protein | |
Research Use Only | |
GCG | |
Recombinant Protein | |
E. coli | |
HSQGTFTSDY SKYLDSRRAQ DFVQWLMNTK RNRNNIA | |
Greater than 90% as determined by SDS-PAGE. |
2641 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |