missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucagon Protein
A DNA sequence encoding the Homo sapiens (Human) Glucagon, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_002045.1 |
---|---|
Gene ID (Entrez) | 2641 |
Name | Glucagon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065701
|
enQuireBio™
QP8639-ec-10ug |
10 μg | |||||||
16005711
|
enQuireBio™
QP8639-ec-50ug |
50 μg | |||||||
16055701
|
enQuireBio™
QP8639-ec-100ug |
100 μg | |||||||
16085701
|
enQuireBio™
QP8639-ec-200ug |
200 μg | |||||||
16095701
|
enQuireBio™
QP8639-ec-500ug |
500 μg | |||||||
16075701
|
enQuireBio™
QP8639-ec-1mg |
1 mg | |||||||
Specifications
NP_002045.1 | |
Glucagon Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
2641 | |
Research Use Only | |
GCG | |
Recombinant Protein | |
E. coli | |
HSQGTFTSDY SKYLDSRRAQ DFVQWLMNTK RNRNNIA | |
Greater than 90% as determined by SDS-PAGE. |