missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Glycine N-methyltransferase/GNMT
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6526-MT-010
298.91 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Glycine N-methyltransferase/GNMT is derived from E. coli. The Recombinant Human Glycine N-methyltransferase/GNMT has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol. | |
33 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
27232 | |
Glycine N-Methyltransferase/GNMT | |
E. coli-derived human Glycine N-Methyltransferase/GNMT protein Met1-Asp295 | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |