missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycodelin Protein
A DNA sequence encoding the Homo sapiens (Human) Glycodelin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6462-ma-100ug
This item is not returnable.
View return policy
Specifications
NP_001018058.1 | |
Glycodelin Protein | |
Research Use Only | |
PAEP | |
Recombinant Protein | |
Mammal | |
MDIPQTKQDL ELPKLAGTWH SMAMATNNIS LMATLKAPLR VHITSLLPTP EDNLEIVLHR WENNSCVEKK VLGEKTENPK KFKINYTVAN EATLLDTDYD NFLFLCLQDT TTPIQSMMCQ YLARVLVEDD EIMQGFIRAF RPLPRHLWYL LDLKQMEEPC RF | |
Greater than 90% as determined by SDS-PAGE. |
5047 | |
100 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |