missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMPS Protein

A cDNA sequence encoding the GMPS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12008-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939078

  • £5730.00 / 1mg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

GMPS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMALCNG DSKLENAGGD LKDGHHHYEG AVVILDAGAQ YGKVIDRRVR ELFVQSEIFP LETPAFAIKE QGFRAIIISG GPNSVYAEDA PWFDPAIFTI GKPVLGICYG MQMMNKVFGG TVHKKSVRED GVFNISVDNT CSLFRGLQKE EVVLLTHGDS VDKVADGFKV VARSGNIVAG IANESKKLYG AQFHPEVGLT ENGKVILKNF LYDIAGCSGT FTVQNRELEC IREIKERVGT SKVLVLLSGG VDSTVCTALL NRALNQEQVI AVHIDNGFMR KRESQSVEEA LKKLGIQVKV INAAHSFYNG TTTLPISDED RTPRKRISKT LNMTTSPEEK RKIIGDTFVK IANEVIGEMN LKPEEVFLAQ GTLRPDLIES ASLVASGKAE LIKTHHNDTE LIRKLREEGK VIEPLKDFHK DEVRILGREL GLPEELVSRH PFPGPGLAIR VICAEEPYIC KDFPETNNIL KIVADFSASV KKPHTLLQRV KACTTEEDQE KLMQITSLHS LNAFLLPIKT VGVQGDCRSY SYVCGISSKD EPDWESLIFL ARLIPRMCHN VNRVVYIFGP PVKEPPTDVT PTFLTTGVLS TLRQADFEAH NILRESGYAG KISQMPVILT PLHFDRDPLQ KQPSCQRSVV IRTFITSDFM TGIPATPGNE IPVEVVLKMV TEIKKIPGIS RIMYDLTSKP PGTTEWE
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GMPS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers