missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GORASP2 Protein
A cDNA sequence encoding the GORASP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12026-5ug
This item is not returnable.
View return policy
Specifications
GORASP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGSSQSV EIPGGGTEGY HVLRVQENSP GHRAGLEPFF DFIVSINGSR LNKDNDTLKD LLKANVEKPV KMLIYSSKTL ELRETSVTPS NLWGGQGLLG VSIRFCSFDG ANENVWHVLE VESNSPAALA GLRPHSDYII GADTVMNESE DLFSLIETHE AKPLKLYVYN TDTDNCREVI ITPNSAWGGE GSLGCGIGYG YLHRIPTRPF EEGKKISLPG QMAGTPITPL KDGFTEVQLS SVNPPSLSPP GTTGIEQSLT GLSISSTPPA VSSVLSTGVP TVPLLPPQVN QSLTSVPPMN PATTLPGLMP LPAGLPNLPN LNLNLPAPHI MPGVGLPELV NPGLPPLPSM PPRNLPGIAP LPLPSEFLPS FPLVPESSSA ASSGELLSSL PPTSNAPSDP ATTTAKADAA SSLTVDVTPP TAKAPTTVED RVGDSTPVSE KPVSAAVDAN ASESP | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GORASP2 solution (1 mg/1ml) contains Phosphate buffered saline (pH 7.4). |