missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Group XIIA secretory phospholipase A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Group XIIA secretory phospholipase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9890-ye-100ug
This item is not returnable.
View return policy
Specifications
NP_110448.2 | |
Group XIIA secretory phospholipase A2 Protein | |
Research Use Only | |
PLA2G12A | |
Recombinant Protein | |
Yeast | |
QEQAQTTDWR ATLKTIRNGV HKIDTYLNAA LDLLGGEDGL CQYKCSDGSK PFPRYGYKPS PPNGCGSPLF GVHLNIGIPS LTKCCNQHDR CYETCGKSKN DCDEEFQYCL SKICRDVQKT LGLTQHVQAC ETTVELLFDS VIHLGCKPYL DSQRAACRCH YEE | |
Greater than 90% as determined by SDS-PAGE. |
81579 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |