missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GSTO1 Protein
A cDNA sequence encoding the GSTO1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £4955.00
Specifications
Name | GSTO1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949238
|
enQuireBio™
QP12061-HIS-EC-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15929238
|
enQuireBio™
QP12061-HIS-EC-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15939238
|
enQuireBio™
QP12061-HIS-EC-1mg |
1 mg |
£4955.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
GSTO1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GSTO1 protein is supplied in 20 mM Tris pH 8.0 and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGESARSLG KGSAPPGPVP EGSIRIYSMR FCPFAERTRL VLKAKGIRHE VININLKNKP EWFFKKNPFG LVPVLENSQG QLIYESAITC EYLDEAYPGK KLLPDDPYEK ACQKMILELF SKVPSLVGSF IRSQNKEDYA GLKEEFRKEF TKLEEVLTNK KTTFFGGNSI SMIDYLIWPW FERLEAMKLN ECVDHTPKLK LWMAAMKEDP TVSALLTSEK DWQGFLELYL QNSPEACDYG L | |
Greater than 95.0% as determined by SDS-PAGE. |