missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HB-EGF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
HB-EGF protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1 ng/mL, corresponding to a specific activity of >1.0 x 106IU/mg.
Specifications
Specifications
| Accession Number | Q99075, Q99075, Q99075 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 10 mM PBS, 500 mM NaCl, pH 7.4. |
| Gene ID (Entrez) | 1839 |
| Molecular Weight (g/mol) | 9.7 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of HB-EGF as determined by LAL method. |
| Gene Alias | diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction