Learn More
enQuireBio™ Recombinant Human HBEGF / DTR Protein
Recombinant Protein
Brand:  enQuireBio™ QP5305-10ug
 View more versions of this product
Product Code. 15935889
Specifications
NP_001936.1
HBEGF / DTR Protein
Research Use Only
HBEGF
Recombinant Protein
E. coli
MDLQEADLDL . LRVTLSSKP Q. ALATPNKE EH. GKRKKKG KGL. GKKRDP CLRK. YKDFC IHGEC. KYVK ELRAPS. CIC HPGYHGERCH GLSL
Greater than 95% as determined by SDS-PAGE.
1839
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
3T3 cell proliferation is significantly induced by less than 1 ng/ml Human HBEGF / DTR, (activity greater than 1.0 x 10^6 units/mg).
Human
Untagged
10 mM sodium phosphate, pH 7.5