missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HEXIM1 Protein
A cDNA sequence encoding the HEXIM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12226-2ug
This item is not returnable.
View return policy
Specifications
HEXIM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMAE PFLSEYQHQP QTSNCTGAAA VQEELNPERP PGAEERVPEE DSRWQSRAFP QLGGRPGPEG EGSLESQPPP LQTQACPESS CLREGEKGQN GDDSSAGGDF PPPAEVEPTP EAELLAQPCH DSEASKLGAP AAGGEEEWGQ QQRQLGKKKH RRRPSKKKRH WKPYYKLTWE EKKKFDEKQS LRASRIRAEM FAKGQPVAPY NTTQFLMDDH DQEEPDLKTG LYSKRAAAKS DDTSDDDFME EGGEEDGGSD GMGGDGSEFL QRDFSETYER YHTESLQNMS KQELIKEYLE LEKCLSRMED ENNRLRLESK RLGGDDARVR ELELELDRLR AENLQLLTEN ELHRQQERAP LSKFGD | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HEXIM1 solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |