missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGFR/c-MET Fc Chimera His-tag Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 358-MT-100/CF
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human HGF R/c-MET Fc Chimera His-tag Protein is derived from NS0. The Recombinant Human HGF R/c-MET Fc Chimera His-tag Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
32.5 kDa (alpha chain), 96.7 kDa (beta chain) | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
Recombinant |
4233 | |
HGFR/c-MET | |
Mouse myeloma cell line,NS0-derived human HGF R/c-MET protein, (N-terminus) HGF R alpha (Glu25-Arg307) Accession &Num; P08581 HGF R beta (Ser308-Thr932) Accession &Num; P08581 HIEGRMD Human IgG1 (Pro100-Lys330) 6 His tag (C-terminus) | |
Tetramer containing two disulfide-linked heterodimers (proteolytically cleaved alpha and beta subunits) | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |