missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human His6-USP1/His6-UAF1 Complex Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ E-568-050

353.34 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18146680

  • £462.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recombinant Human USP1/UAF1 complex is a Ubiquitin-specific deconjugating enzyme heterodimer. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP1/UAF1 complex concentration of 0.01-0.5 μM.
Specifications

Specifications

O94782.1 (USP1), Q8TAF3.1 (UAF1)
Supplied as a 0.2 μm filtered solution in HEPES, NaCl, TCEP and Glycerol.
M.W. (theoretical): 89 kDa (USP1), 77 kDa (UAF1)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Unconjugated
>80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Bioactivity
7398
His6 USP1/UAF1 Complex
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP1/UAF1 Complex protein Met1 - Leu785 (Gly670Ala, Gly671Ala) with a C-terminal 6-His tag (USP1), Met1 - Thr677 with a C-terminal 6-His tag (UAF1)
USP1
Recombinant
USP1/UAF1 Complex
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers