missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Histamine N-Methyltransferase/HNMT

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 7637-MT-010

261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18357767

This item is not returnable. View return policy

Description

Description

The Recombinant Human Histamine N-Methyltransferase/HNMT is derived from E. coli. The Recombinant Human Histamine N-Methyltransferase/HNMT has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol.
34 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
3176
Histamine N-Methyltransferase/HNMT
E. coli-derived human Histamine N-Methyltransferase/HNMT protein Ala2-Ala292,with an N-terminal HM and 6-His tag.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers