missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8907-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091886.1 | |
Histone deacetylase 7 Protein | |
Research Use Only | |
HDAC7 | |
Recombinant Protein | |
E. coli | |
VTDLAFKVAS RELKNGFAVV RPPGHHADHS TAMGFCFFNS VAIACRQLQQ QSKASKILIV DWDVHHGNGT QQTFYQDPSV LYISLHRHDD GNFFPGSGAV DEVGAGSGEG FNVNVAWAGG LDPPMGDPEY LAAFRIVVMP IAREFSPDLV LVSAGFDAAE GHPAPLGGYH VSAKCFGYMT QQLMNLAGGA VVLALEGGHD LTAICDASEA CVAALLGNRV DPLSEEGWKQ KPNLNAIRSL EAVIRVHSKY WGCMQRLASC PDSWVPRVPG A | |
Greater than 90% as determined by SDS-PAGE. |
51564 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |