missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HLA DQB1 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00003119-Q01-25ug
490.40 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification | |
3119 | |
25 μg | |
DC-1 alpha chain, DC-alpha, HLA class II histocompatibility antigen, DQ alpha 1 chain-like, HLA-DCA, MHC class II DQA1 | |
Asthma, Diabetes Research, Immunology | |
HLA DQB1 |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
35.42 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
HLA-DQB1 | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction