missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSF2BP Protein

A cDNA sequence encoding the HSF2BP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12323-1mg

 View more versions of this product

Product Code. 15930059

  • £2975.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HSF2BP Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGEAGAA EEACRHMGTK EEFVKVRKKD LERLTTEVMQ IRDFLPRILN GEVLESFQKL KIVEKNLERK EQELEQLKMD CEHFKARLET VQADNIREKK EKLALRQQLN EAKQQLLQQA EYCTEMGAAA CTLLWGVSSS EEVVKAILGG DKALKFFSIT GQTMESFVKS LDGDVQELDS DESQFVFALA GIVTNVAAIA CGREFLVNSS RVLLDTILQL LGDLKPGQCT KLKVLMLMSL YNVSINLKGL KYISESPGFI PLLWWLLSDP DAEVCLHVLR LVQSVVLEPE VFSKSASEFR SSLPLQRILA MSKSRNPRLQ TAAQELLEDL RTLEHNV
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSF2BP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 30% glycerol and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers